Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 16 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
£128.00 - £185.00
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12408230
|
Tenak™
TE15458 |
Microtiterplates (115) |
£185.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12418210
|
Tenak™
TE15404 |
Microtiterplates (64) |
£152.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
12418250
|
Tenak™
TE15520 |
Microtiterplates (54) |
£144.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12428210
|
Tenak™
TE15406 |
Microtiterplates (68) |
£164.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
12428270
|
Tenak™
TE15570 |
Microtiterplates (63) |
£148.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12438250
|
Tenak™
TE15524 |
Microtiterplates (96) |
£169.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
12448250
|
Tenak™
TE15526 |
Microtiterplates (114) |
£181.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
12458270
|
Tenak™
TE15576 |
Microtiterplates (133) |
£185.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
12468220
|
Tenak™
TE15450 |
Microtiterplates (45) |
£144.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12488220
|
Tenak™
TE15454 |
Microtiterplates (80) |
£160.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
12498200
|
Tenak™
TE15400 |
Microtiterplates (36) |
£128.00
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
12498220
|
Tenak™
TE15456 |
Microtiterplates (95) |
£172.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum