missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 16 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Brand: Tenak™ TE15458
Additional Details : Weight : 1.60000kg
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
23 x 5 | |
Aluminum | |
450 x 135 x 391 mm |
Microtiterplates (115) | |
Upright Freezers | |
16 mm |