missing translation for 'onlineSavingsMsg'
Learn More

ECM1 Rabbit, anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen ECM1
Concentration 450 μg/mL
Applications Western Blot, ELISA
Form Liquid
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18007305
View Documents
Proteintech
30725-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18087075
View Documents
Proteintech
30725-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ECM1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ECM1. This antibody recognizes human antigen. The ECM1 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

ECM1
Western Blot, ELISA
Rabbit
1893
IgG
Store at -20°C. Stable for one year after shipment.
450 μg/mL
Liquid
Human
ECM1, extracellular matrix protein 1, Secretory component p85
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.