Learn More
ECM1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30725-1-AP-20UL
This item is not returnable.
View return policy
Description
The ECM1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ECM1. This antibody recognizes human antigen. The ECM1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ECM1 | |
| 450 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| ECM1, extracellular matrix protein 1, Secretory component p85 | |
| 20 μL | |
| 1893 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |