missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Elma Clean 145 Cleaning Concentrate

Product Code. 11711369 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
25 L
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11711369 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11711369

Please to purchase this item. Need a web account? Register with us today!

Suitable for the slightly acid cleaning of nonferrous heavy metals, aluminum, stainless steel, glass, plastics and alloys. Fisherbrand™ Elma Clean 145 Cleaning Concentrate is also good at decalcifying.

  • Has an anti-static effect
  • Results considerably improved by pre-cleaning with an alkaline cleaner
  • Removes polishing media, grease, oil, oxide layers and lime
  • Risk of tarnish when different metals are cleaned in the same bath

Specifications

Form Liquid
pH 1
Appearance Light Brown
Boiling Point ≥100°C
Composition Anionic surfactants 5 to 15%, non-ionic surfactants 5 to 15%
Concentration Ultrasonic bath 2 to 5%
Container Type Can
Content And Storage 5°C to 30°C
Volume (Metric) 25 L
compliance-icons compliance-icons
Product Identifier
  • elma clean 145 (EC 145)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: cocosfattyaminoxethylate, alkylbenzenesulphonic acid, tartaric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.