missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBV Latent Membrane Protein 1 Antibody (LMPO24) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £328.00
Specifications
| Antigen | EBV Latent Membrane Protein 1 |
|---|---|
| Clone | LMPO24 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Flow (Intracellular) 1:10 - 1:1000, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663579
|
Novus Biologicals
NBP2-50383-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606636
|
Novus Biologicals
NBP2-50383 |
0.1 mL |
£328.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EBV Latent Membrane Protein 1 Monoclonal antibody specifically detects EBV Latent Membrane Protein 1 in Virus samples. It is validated for Flow Cytometry, Immunohistochemistry, Flow (Intracellular), CyTOFSpecifications
| EBV Latent Membrane Protein 1 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Signaling, Immunology, Virology Bacteria and Parasites | |
| PBS | |
| IgG1 κ | |
| Protein A purified |
| LMPO24 | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Flow (Intracellular) 1:10 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Purified plasma membranes from EBV Latent Membrane Protein 1 expressing insect cells infected with a recombinant EBV Latent Membrane Protein 1 baculovirus | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title