missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBV Latent Membrane Protein 1 Antibody (LMPO24) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50383-0.025ml
This item is not returnable.
View return policy
Description
EBV Latent Membrane Protein 1 Monoclonal antibody specifically detects EBV Latent Membrane Protein 1 in Virus samples. It is validated for Flow Cytometry, Immunohistochemistry, Flow (Intracellular), CyTOF
Specifications
| EBV Latent Membrane Protein 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Flow (Intracellular) 1:10 - 1:1000, CyTOF-ready | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| LMPO24 | |
| Unconjugated | |
| PBS | |
| Purified plasma membranes from EBV Latent Membrane Protein 1 expressing insect cells infected with a recombinant EBV Latent Membrane Protein 1 baculovirus | |
| 0.025 mL | |
| Cancer, Cellular Signaling, Immunology, Virology Bacteria and Parasites | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction