missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Annexin V Apoptosis Detection Kits

Product Code. p-7236177
Click to view available options
Quantity:
100 Tests
20 Tests
200 Test
200 Tests
300 Tests
50 Tests
Excitation/Emission:
405, 546/450, 647
488, 535/520, 617
650, 535/660, 617
NA, 535/NA, 617
Conjugate:
APC, Propidium Iodide
Biotin, Propidium Iodide
FITC, Propidium Iodide
PCP-eFluor 710
PE, 7-AAD
PE/Cy7
eFluor 450, 7-AAD
Förpackningsstorlek:
100 tests
20 tests
200 tests
300 tests
50 tests
Each
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 12770170

Brand: Invitrogen™ 88800574

Please to purchase this item. Need a web account? Register with us today!

Edge
Denna artikel kan inte returneras. Se returpolicy

Innehåller

  • Cat. No. 88-8005-72:
  • 10X Binding Buffer
  • Annexin V FITC : 5μL/test, 50 tests Protect from light.
  • Propidium Iodide Staining Solution : 5μL/test, 100 tests Protect from light.
  • Cat. No. 88-8005-74:
  • 10X Binding Buffer
  • Annexin V FITC : 5μL/test, 200 tests Protect from light.
  • Propidium Iodide Staining Solution : 5μL/test, 2x100 tests Protect from light.

Kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane.

Easily distinguish cells undergoing early and late apoptosis with eBioscience and Annexin V Apoptosis Detection kits for flow cytometry analysis. These kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane. In late-stage apoptosis, as cell integrity is lost, both annexin V and the viability dye traverse and bind PS on the interior of the plasma membrane.

Annexins are a family of calcium-dependent phospholipid-binding proteins that preferentially bind phosphatidylserine (PS). Under normal physiologic conditions, PS is predominantly located in the inner leaflet of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution across the phospholipid bilayer and is translocated to the extracellular membrane leaflet marking cells as targets of phagocytosis. Once on the outer surface of the membrane, PS can be detected by fluorescently labeled annexin V in a calcium-dependent manner.

In early-stage apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI), 7-AAD, or fixable viability dyes such as eFluor 450. These cells will stain with annexin V but not a viability dye, thus distinguishing cells in early apoptosis. However, in late-stage apoptosis, the cell membrane loses integrity, thereby allowing annexin V to also access PS in the interior of the cell. A viability dye can be used to resolve these late-stage apoptotic and necrotic cells (annexin V, viability dye-positive) from the early-stage apoptotic cells (annexin V positive, viability dye-negative).

Current viability stains available with the eBioscience Annexin V Apoptosis Detection kits include FITC, eFluor 450, allophycocyanin (APC), phycoerythrin (PE), PE-cyanine7, and biotin.

TRUSTED_SUSTAINABILITY

Specifikationer

Description eBioscience Annexin V Apoptosis Detection Kit FITC
Quantity 200 Tests
Format Kit
Kit Contents • Annexin V Binding Buffer (10X), 1 bottle (100 mL)
• Annexin V-FITC conjugate, 1 vial (1 mL)
• Propidium Iodide Staining Solution, 2 vials (0.5 mL)

Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits.
Product Type Apoptosis Detection Kit
Content And Storage 2°C to 8°C. Avoid temperature fluctuations.
Excitation/Emission 488, 535/520, 617
Product Line eBioscience
Conjugate FITC, Propidium Iodide
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.