missing translation for 'onlineSavingsMsg'
Learn More

Ebastine N-Oxide, TRC

Product Code. 30009827
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009827 10 mg 10mg
30006481 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009827 Supplier Toronto Research Chemicals Supplier No. TRCE32001510MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256285-71-8
Molecular Formula C32 H39 N O3
Molecular Weight (g/mol) 485.66
Synonym Ebastine N-Oxide (cis and trans), 1-Butanone, 1-[4-(1,1-dimethylethyl)phenyl]-4-[4-(diphenylmethoxy)-1-oxido-1-piperidinyl]-
IUPAC Name 4-(4-benzhydryloxy-1-oxidopiperidin-1-ium-1-yl)-1-(4-tert-butylphenyl)butan-1-one
SMILES CC(C)(C)c1ccc(cc1)C(=O)CCC[N+]2([O-])CCC(CC2)OC(c3ccccc3)c4ccccc4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C32H39NO3/c1-32(2,3)28-18-16-25(17-19-28)30(34)15-10-22-33(35)23-20-29(21-24-33)36-31(26-11-6-4-7-12-26)27-13-8-5-9-14-27/h4-9,11-14,16-19,29,31H,10,15,20-24H2,1-3H3
Formula Weight 485.293
Chemical Name or Material Ebastine N-Oxide (cis and trans)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.