CAS RN 1256285-71-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ebastine N-Oxide, TRC
CAS: 1256285-71-8 Molecular Formula: C32 H39 N O3 Molecular Weight (g/mol): 485.66 Synonym: Ebastine N-Oxide (cis and trans),1-Butanone, 1-[4-(1,1-dimethylethyl)phenyl]-4-[4-(diphenylmethoxy)-1-oxido-1-piperidinyl]- IUPAC Name: 4-(4-benzhydryloxy-1-oxidopiperidin-1-ium-1-yl)-1-(4-tert-butylphenyl)butan-1-one SMILES: CC(C)(C)c1ccc(cc1)C(=O)CCC[N+]2([O-])CCC(CC2)OC(c3ccccc3)c4ccccc4
2
Ebastine N-Oxide (cis and trans), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.