missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00342-50ul
This item is not returnable.
View return policy
Description
EB3 Polyclonal antibody specifically detects EB3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| EB3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| EB1 protein family member 3, EB3EBF3-S, End-binding protein 3, microtubule-associated protein RP/EB family member 3, microtubule-associated protein, RP/EB family, member 3, RP3EBF3APC binding protein | |
| Produced in rabbits immunized with purified, recombinant Human EB3 (Uniprot#: Q9UPY8-1; Met1-Tyr281) | |
| 50 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, GPCR, Signal Transduction | |
| 22924 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction