missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£180.00 - £248.00
Specifications
| Antigen | EB3 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:1000-1:4000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684688
|
Novus Biologicals
NBP3-00342-100ul |
100 μg |
£248.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639337
|
Novus Biologicals
NBP3-00342-50ul |
50 μg |
£180.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EB3 Polyclonal antibody specifically detects EB3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| EB3 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, GPCR, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 22924 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EB1 protein family member 3, EB3EBF3-S, End-binding protein 3, microtubule-associated protein RP/EB family member 3, microtubule-associated protein, RP/EB family, member 3, RP3EBF3APC binding protein | |
| Produced in rabbits immunized with purified, recombinant Human EB3 (Uniprot#: Q9UPY8-1; Met1-Tyr281) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title