missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Complete reactor system for organic or aqueous syntheses in the laboratory
- Can be controlled with IKA labworldsoft™ 6 lab software (available as an accessory) to automate all processes
- High-quality materials allow operation at temperatures from -50°C to +200°C and up to a vacuum of 3 mbar
- LT 5.40 draining set with special valves enables fast, clean draining of the tempering fluid from the double jacket and easy replacement of the reactor vessel
Includes
- Ergonomically designed stand system for quick and easy replacement of reactor vessels and adjustment of the working height
- DN 100 (500 mL and 1000 mL) or DN 150 (2000 mL and 5000 mL) vessel clamp for holding the reactor vessel
- Double-walled reaction vessel made of chemically resistant borosilicate 3.3 glass with bottom discharge valve
- Spring-loaded valve spindle of the drain valve to prevent glass breakage
- Reactor lid with various openings (1 x NS 29/32 center glass joint, 3 x NS 29/32, 1 x NS 14/23, 2 x GL 18) for flexible adaptation to the desired application
- DN 100 or DN 150 clamp ring and FEP-coated O-ring to ensure chemical resistance
- EUROSTAR 100 control overhead stirrer with 200 Ncm, keyless chuck, high-precision torque monitoring (±1 Ncm accuracy) and viscosity measurement (mPas)
- Timer, overload protection and temperature measurement integrated in the overhead stirrer
- PTFE-coated propeller stirrer for optimum mixing and chemical resistance
- High-quality stirrer guide for vacuum operation up to 3 mbar
- LT 5.40 drainage set with valves and temperature control hoses
- RS 232, Ethernet, USB or wireless connection interfaces on the stirrer for external control with labworldsoft™ 6
Specifications
Specifications
| Volume (Metric) | 1000 mL |
| Product Type | Lab Reactor |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?