missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Release Human PSC-Derived Neural Crest Cell Positive Selection Kit

Product Code. 17117461
Encompass
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17117461

Brand: STEMCELL Technologies 1000047

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic positive selection kit using particle release technology

  • The EasySep™ Release Human PSC-Derived Neural Crest Cell Positive Selection Kit is designed to isolate CD271+ cells from human pluripotent stem cell (PSC)-derived neural crest cell culture by immunomagnetic positive selection
  • Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
  • Unwanted cells are simply poured off, while desired cells remain in the tube
  • Then, bound magnetic particles are removed from the EasySep™-isolated CD271+ cells, which are immediately available for downstream applications
  • Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands

Specifications

Cell Type Neural Cells, PSC-Derived
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.