missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ RBC Depletion Reagent - 10mL

Product Code. 17177851
Encompass
Change view
Click to view available options
Quantity:
100 mL Whole Blood
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17177851 100 mL Whole Blood 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17177851 Supplier STEMCELL Technologies Supplier No. 18170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic isolation of untouched human leukocytes from whole blood

  • The EasySep™ RBC Depletion Reagent is designed to immunomagnetically deplete red blood cells (RBCs) from fresh human peripheral blood, umbilical cord blood, bone marrow or leukapheresis samples by negative selection
  • RBCs are targeted with Tetrameric Antibody Complexes recognizing Glycophorin A and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Unwanted cells remain in the tube while desired cells are poured off
  • This product replaces the EasySep™ Human Glycophorin A Depletion Kit (Catalog #18352) for even faster cell isolations

Specifications

Quantity 100 mL Whole Blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.