missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Mouse Mesenchymal Stem/Progenitor Cell Enrichment Kit
Description
- The EasySep™ Mouse Mesenchymal Stem/Progenitor Cell Enrichment Kit is designed to isolate mesenchymal stem/progenitor cells from mouse compact bone by negative selection
- Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-mesenchymal stem/progenitor cells (CD45, TER119)
- Labeled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin and dextran
- These cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
Specifications
| Cell Type | Mesenchymal Stem and Progenitor Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 4 x 108 Cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?