missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Mouse Hematopoietic Progenitor Cell Isolation Kit
Description
- The EasySep™ Mouse Hematopoietic Cell Isolation Kit is designed to isolate stem and progenitor cells from single-cell suspensions of bone marrow or other tissues by negative selection
- Unwanted cells are targeted for removal with biotinylated antibodies directed against non-hematopoietic stem cells and non-progenitor cells (CD5, CD11b, CD19, CD45R/B220, Ly6G/C(Gr-1), TER119, 7-4) and streptavidin-coated magnetic particles (RapidSpheres™ )
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
- This product replaces the EasySep™ Mouse Hematopoietic Progenitor Cell Enrichment Kit (Catalog #19756) for faster cell isolations
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?