Learn More
STEMCELL Technologies EasySep™ Mouse Epithelial Cell Enrichment Kit II
Immunomagnetic isolation of untouched mouse epithelial cells
Brand: STEMCELL Technologies 19868
Additional Details : Weight : 0.03200kg
Description
- The EasySep™ Mouse Epithelial Cell Enrichment Kit II is designed to isolate epithelial cells from freshly dissociated mouse mammary tissues by immunomagnetic negative selection
- The EasySep™ procedure involves labeling unwanted non-epithelial cells with biotinylated antibodies and magnetic particles
- The magnetically labeled cells are separated from untouched desired cells by using an EasySep™ magnet and simply pouring the desired cells into a new tube
- This product replaces the EasySep™ Epithelial Cell Enrichment Kit (Catalog #19758) for even faster isolations
- It also replaces the cell enrichment portion of the EasySep™ Mouse Mammary Stem Cell Enrichment Kit (Catalog #19757)
- Additionally, Anti-Mouse CD24 Antibody, Clone M1/69, PE (Catalog #60099PE) and Anti-Mouse CD49f Antibody, Clone GoH3, FITC (Catalog #60037FI) are required to complete the kit
Specifications
Mammary Cells, Prostate Cells | |
Cell Isolation | |
1 x 109 cells |