missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Mouse CD45 Positive Selection Kit
Description
- The EasySep™ Mouse CD45 Positive Selection Kit is designed to isolate CD45 cells from single-cell suspensions of splenocytes, lung, or other tissues by positive selection
- Desired cells are targeted with antibody complexes recognizing CD45 and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
Specifications
Specifications
| Cell Type | B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Innate Lymphoid Cells, Leukemia/Lymphoma Cells, Lymphocytes, Macrophages, Megakaryocytes, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Plasma, T Cells, T Cells, Other Subsets |
| For Use With (Application) | Cell Isolation |
| Quantity | 2 x 109 cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?