missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human NK Cell Isolation Kit

Product Code. 16884213
Encompass
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16884213

Brand: STEMCELL Technologies 17955

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic isolation of untouched human NK cells

  • The EasySep™ Human NK Cell Isolation Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
  • This product can be used in place of the EasySep™ Human NK Cell Enrichment Kit (Catalog #19055) for even faster cell isolations

Specifications

Cell Type NK Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.