missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human Neutrophil Isolation Kit

Product Code. 17107831
Encompass
Change view
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17107831 1 x 109 cells 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17107831 Supplier STEMCELL Technologies Supplier No. 17957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic isolation of untouched human neutrophils

  • The EasySep™ Human Neutrophil Isolation Kit is designed to isolate neutrophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-neutrophils and magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
  • The isolation kit is compatible with cells prepared using HetaSep™ (Catalog #07906) sedimentation or red blood cell lysis
  • This product replaces the EasySep™ Human Neutrophil Enrichment Kit (Catalog #19257), for even faster cell isolations

Specifications

Cell Type Granulocytes and Subsets
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.