missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human Monocyte Enrichment Kit
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
300mL
Description
- The EasySep™ Human Monocyte Enrichment Kit is designed to isolate monocytes from fresh or previously frozen peripheral blood mononuclear cells by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-monocyte cells and dextran-coated magnetic particles
- The cocktail also contains an antibody to human Fc receptor to prevent nonspecific binding of monocytes
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
- For applications in which CD16+ cells are not removed, we recommend the EasySep™ Human Monocyte Enrichment Kit without CD16 Depletion (Catalog #19058)
Specifications
Specifications
| Cell Type | Monocytes |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction