missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human Memory CD4+ T Cell Enrichment Kit
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
Each
Description
- The EasySep™ Human Memory CD4 T Cell Enrichment Kit is designed to isolate memory CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD8, CD14, CD16, CD19, CD20, CD36, CD45RA, CD56, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles
- The labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
Specifications
| Cell Type | T Cells, T Cells, CD4+ |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction