missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human IgG+ Memory B Cell Isolation Kit

Product Code. 17177771
Encompass
Change view
Click to view available options
:
For Cell Isolation
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17177771 For Cell Isolation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17177771 Supplier STEMCELL Technologies Supplier No. 17868

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Immunomagnetic cell isolation kit using particle release technology

  • The EasySep™ Human IgG+ Memory B Cell Isolation Kit isolates IgG+CD27+ B cells from fresh or previously frozen peripheral blood mononuclear cells (PBMCs) or leukapheresis samples
  • First, CD27+ cells are isolated by column-free immunomagnetic positive selection using EasySep™ Releasable RapidSpheres™
  • Then, bound magnetic particles are removed from the EasySep™-isolated CD27+ cells, and unwanted non-B cells, as well as IgM+, IgD+ and IgA+ cells are depleted using EasySep™ Dextran RapidSpheres™
  • The final isolated fraction contains IgG+CD27+ memory B cells, which readily secrete IgG in culture supernatants in the presence of appropriate stimuli

Specifications

Cell Type B Cells
For Use With (Application) Cell Isolation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.