missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human FITC Positive Selection Kit II
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Description
- The EasySep™ Human FITC Positive Selection Kit II is designed to isolate cells that are labeled with FITC-conjugated antibodies
- Desired cells are targeted with antibody complexes recognizing FITC and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- This product replaces the EasySep™ Human FITC Positive Selection Kit (Catalog #18552)
Specifications
Specifications
| Cell Type | B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction