missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human Central and Effector Memory CD4+ T Cell Isolation Kit
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Description
- This kit isolates human CD62L+CD45RO+ and CD62L-CD45RO+ memory CD4+ T cells from fresh peripheral blood mononuclear cells (PBMCs) or washed leukapheresis samples
- First, CD62L+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™
- Then, bound magnetic particles are removed from the EasySep™-isolated CD62L+ cells, and unwanted CD45RA+ and non-CD4+ T cells are targeted for depletion using antibody complexes and EasySep™ Dextran RapidSpheres™
- The final isolated fraction contains highly purified CD62L+CD45RO+ central memory CD4+ T cells that are immediately ready for downstream applications
- Effector memory CD4+ T cells can be isolated from the CD62L- fraction
Specifications
Specifications
| Cell Type | T Cells, CD4+ |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction