missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human CD4+CD127lowCD25+ Regulatory T Cell Isolation Kit, with RoboSep™ Buffer and Filter Tips
Description
- This kit isolates human CD4+CD127lowCD25+ regulatory T cells (Tregs) from fresh peripheral blood mononuclear cells (PBMCs) or leukapheresis samples
- First, CD25+ cells are isolated by column-free immunomagnetic positive selection using EasySep™ Releasable RapidSpheres™
- Then, bound magnetic particles are removed from the EasySep™-isolated CD25+ cells, and unwanted non-Tregs are targeted for depletion
- The final isolated fraction contains highly purified CD4+CD127lowCD25+ cells that express high levels of FOXP3 and are immediately ready for downstream applications
- An optional protocol allows for the isolation of CD4+CD25- responder T cells in parallel for use in functional studies
- Following cell isolation with this EasySep™ kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins, or other chemically related ligands
Specifications
Specifications
| Cell Type | T Cells, T Cells, CD4+, T Cells, Regulatory |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?