missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human CD34 Positive Selection Kit II

Product Code. 17137771
Encompass
Click to view available options
Quantity:
5 x 109 Cells
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17137771

Brand: STEMCELL Technologies 17856

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic positive selection of human CD34+ cells (e.g. hematopoietic, stem/progenitor cells)

  • The EasySep™ Human CD34 Positive Selection Kit II is designed to isolate CD34+ cells from fresh or previously frozen bone marrow or mobilized peripheral blood mononuclear cells, or from previously frozen cord blood mononuclear cells, or from hESC/hiPSC-derived hematopoietic cells by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD34 and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD34 antigen is expressed on hematopoietic stem and progenitor cells
  • This product can be used in place of the EasySep™ Human CD34 Positive Selection Kit (Catalog #18056) for even faster cell isolations

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells, Pluripotent Stem Cells
For Use With (Application) Cell Isolation
Quantity 5 x 109 Cells
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.