missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human CD19 Positive Selection Kit II
Description
- The EasySep™ Human CD19 Positive Selection Kit II is designed to isolate CD19+ cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection
- Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles
- The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The CD19 antigen is expressed on mature B cells and B cell progenitors but is lost upon maturation to plasma cells
- It is also expressed on follicular dendritic cells
- This product replaces the EasySep™ Human CD19 Positive Selection Kit (Catalog #18054) for even faster cell isolations
Specifications
Specifications
| Cell Type | B Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?