Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD3 Positive Selection Kit
Product Code. 17197771
Encompass
Click to view available options
Quantity:
60 mL Buffy Coat or Whole Blood
Unit Size:
1 set
Product Code. 17197771
Brand: STEMCELL Technologies 17871
Column-free immunomagnetic positive selection of human CD3+ cells for HLA analysis
  • The EasySep™ HLA Chimerism Whole Blood CD3 Positive Selection Kit is designed to isolate CD3+ cells from fresh whole blood or buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD3 and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD3 antigen is expressed on T cells and NKT cells
  • This product replaces the EasySep™ Human Whole Blood CD3 Positive Selection Kit (Catalog #18081) and EasySep™ HLA Whole Blood CD3 Positive Selection Kit (Catalog #18081HLA), for even faster cell isolations
Specifications
Cell Type T Cells
Quantity 60 mL Buffy Coat or Whole Blood