Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit
Click to view available options
Quantity:
60 mL Buffy Coat or Whole Blood
Unit Size:
1 set
Description
- The EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit is designed to isolate CD19+ cells from whole blood or buffy coat by positive selection
- Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles
- The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The CD19 antigen is present on mature B cells and B cell progenitors but is lost upon maturation to plasma cells
- It is also expressed on follicular dendritic cells
- This product replaces the EasySep™ Human Whole Blood CD19 Positive Selection Kit (Catalog #18084) for even faster cell isolations
Specifications
Specifications
| Cell Type | B Cells |
| Quantity | 60 mL Buffy Coat or Whole Blood |