Learn More
STEMCELL Technologies EasySep™ Direct Human PBMC Isolation Kit
Click to view available options
Quantity:
100 mL whole blood
Unit Size:
1 set
Description
- The EasySep™ Direct Human PBMC Isolation Kit is designed to isolate highly purified peripheral blood mononuclear cells (PBMCs) from fresh whole blood, buffy coat, bone marrow, cord blood, or leukapheresis products by negative selection
- Red blood cells (RBCs), platelets, and unwanted cells are targeted for removal with antibody complexes and magnetic particles and separated using an EasySep™ magnet
- Untouched PBMCs are simply collected into a new tube and are immediately available for downstream applications such as flow cytometry, culture, or DNA/RNA extraction
Specifications
Specifications
| Cell Type | B Cells, Lymphocytes, Monocytes, Mononuclear Cells, NK Cells, T Cells |
| Quantity | 100 mL whole blood |