missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Direct Human Basophil Isolation Kit

Product Code. 17137951
Encompass
Change view
Click to view available options
Quantity:
100 mL Whole Blood
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17137951 100 mL Whole Blood 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17137951 Supplier STEMCELL Technologies Supplier No. 19667

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic isolation of basophils directly from whole blood

  • The EasySep™ Direct Human Basophil Isolation Kit isolates functional, highly purified basophils directly from human whole blood by immunomagnetic negative selection
  • No lysis, density gradient centrifugation, or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications

Specifications

Cell Type Granulocytes and Subsets
For Use With (Application) Cell Isolation
Quantity 100 mL Whole Blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.