missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Direct Human B Cell Isolation Kit
Beskrivning
- Isolate highly purified B cells directly from human whole blood without the need for density gradient centrifugation, RBC sedimentation or lysis
- This platform provides fast and fully immunomagnetic negative selection
- This cell isolation kit targets non-B cells for removal with EasySep™ Direct RapidSpheres™ and antibodies recognizing specific surface antigens
- Unwanted cells are separated from desired cells using an EasySep™ magnet, and untouched target cells are then pipetted or poured into a new tube
- Isolated cells can be used directly in downstream applications such as flow cytometry, culture or DNA/RNA extraction
Specifikationer
Specifikationer
| Cell Type | B Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 100 mL whole blood |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?