missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Direct HLA B Cell Isolation Kit for RoboSep™
Description
- The EasySep™ Direct HLA B Cell Isolation Kit can be used to isolate highly purified B cells directly from human whole blood, buffy coat, spleen, or lymph nodes by immunomagnetic negative selection.
- This kit targets non-B cells for removal with antibodies recognizing specific surface markers.
- Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet.
- Desired cells are simply collected into a new tube and can directly be used for downstream applications such as flow cytometry crossmatch.
Specifications
Specifications
| Cell Type | B Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 100 mL Whole Blood |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?