Learn More
STEMCELL Technologies EasySep™ Dead Cell Removal (Annexin V) Kit
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
Each
Description
- The EasySep™ Dead Cell Removal (Annexin V) kit is designed to deplete apoptotic (Annexin V+) cells from cell culture or tissue preparations by immunomagnetic negative selection
- Unwanted cells are labeled with Annexin V, antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are simply poured off into a new tube
- Annexin V binds phosphatidylserine on the outer leaflet of the cell membrane during apoptosis
Specifications
Specifications
| Cell Type | Lymphocytes |
| Quantity | 1 x 109 Cells |