missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAR2/NR2F6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£175.00 - £310.00
Specifications
| Antigen | EAR2/NR2F6 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18177521
|
Novus Biologicals
NBP2-24510SS |
0.025 mg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18120732
|
Novus Biologicals
NBP2-24510 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EAR2/NR2F6 Polyclonal specifically detects EAR2/NR2F6 in Human, Canine, Primate samples. It is validated for Western Blot.Specifications
| EAR2/NR2F6 | |
| Western Blot 0.5-1.0 ug/ml | |
| Polyclonal | |
| Rabbit | |
| GPCR | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 2063 | |
| A portion of amino acids 20-70 of human NR2F6 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Canine, Primate | |
| EAR-2ERBA-related gene-2, EAR2nuclear receptor subfamily 2 group F member 6, ERBAL2v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 2, nuclear receptor subfamily 2, group F, member 6, V-erbA-related protein 2 | |
| NR2F6 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, 92% homologous indog, 85% homologous incow and opossum and 78% homologous in mouse, horseand rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title