missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAR2/NR2F6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24510
This item is not returnable.
View return policy
Description
EAR2/NR2F6 Polyclonal specifically detects EAR2/NR2F6 in Human, Canine, Primate samples. It is validated for Western Blot.
Specifications
| EAR2/NR2F6 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| NR2F6 | |
| A portion of amino acids 20-70 of human NR2F6 was used as the immunogen. | |
| 0.1 mg | |
| GPCR | |
| 2063 | |
| Human, Canine, Primate | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.5-1.0 ug/ml | |
| EAR-2ERBA-related gene-2, EAR2nuclear receptor subfamily 2 group F member 6, ERBAL2v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 2, nuclear receptor subfamily 2, group F, member 6, V-erbA-related protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, 92% homologous indog, 85% homologous incow and opossum and 78% homologous in mouse, horseand rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction