Learn More
Invitrogen™ E6AP Monoclonal Antibody (K29054_11H8), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | E6AP |
|---|---|
| Clone | K29054_11H8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30212038
|
Invitrogen™
MA556175 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30204446
|
Invitrogen™
MA555795 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: LQAKDEDKDE DEKEKAACSA AAMEEDSEAS SSRIGDSSQG DNNLQKLGPD DVSVDIDAIR RVYTRLLSNE KIETAFLNAL VYLSPNVECD LTYHNVYSRD PNYLNLFIIV MENRNLHSPE YLEMALPLFC KAMSKLPLAA QGKLIRLWSK YNADQIRRMM ETFQQLITYK VISNEFNSRN LVNDDDAIVA ASKCLKMVYY ANVVGGEVDT NHNEEDDEEP IPESSELTLQ ELLGEERRNK KGPRVDPLET ELGVKTLDCR KPLIPFEEFI NEPLNEVLEM DKDYTFFKVE TENKFSFMTC PFILNAVTKN LGLYYDNRIR MYSERRITVL YSLVQGQQLN PYLRLKVRRD HIIDDALVRL EMIAMENPAD LKKQLYVEFE GEQGVDEGGV SKEFFQLVVE EIFNPDIGMF TYDESTKLFW FNPSSFETEG
E6AP Monoclonal antibody specifically detects E6AP in Human samples. It is validated for Western BlotSpecifications
| E6AP | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 4732496B02; 5830462N02Rik; A130086L21Rik; ANCR; AS; CTCL tumor antigen se37-2; E6AP; E6-AP; E6-AP ubiquitin protein ligase; E6AP ubiquitin-protein ligase; EPVE6AP; FLJ26981; HECT-type ubiquitin transferase E3A; HPVE6A; human papilloma virus E6-associated protein; human papillomavirus E6-associated protein; mKIAA4216; oncogenic protein-associated protein E6-AP; Renal carcinoma antigen NY-REN-54; UBE3A; ubiquitin conjugating enzyme E3A; ubiquitin protein ligase E3A; ubiquitin-protein ligase E3A | |
| Recombinant human UBE3A (immunogen range is 181-610). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| UBE3A |
| K29054_11H8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q05086 | |
| 7337 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.