Learn More
Invitrogen™ E6AP Monoclonal Antibody (K29054_11H8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556175
Description
Sequence of this protein is as follows: LQAKDEDKDE DEKEKAACSA AAMEEDSEAS SSRIGDSSQG DNNLQKLGPD DVSVDIDAIR RVYTRLLSNE KIETAFLNAL VYLSPNVECD LTYHNVYSRD PNYLNLFIIV MENRNLHSPE YLEMALPLFC KAMSKLPLAA QGKLIRLWSK YNADQIRRMM ETFQQLITYK VISNEFNSRN LVNDDDAIVA ASKCLKMVYY ANVVGGEVDT NHNEEDDEEP IPESSELTLQ ELLGEERRNK KGPRVDPLET ELGVKTLDCR KPLIPFEEFI NEPLNEVLEM DKDYTFFKVE TENKFSFMTC PFILNAVTKN LGLYYDNRIR MYSERRITVL YSLVQGQQLN PYLRLKVRRD HIIDDALVRL EMIAMENPAD LKKQLYVEFE GEQGVDEGGV SKEFFQLVVE EIFNPDIGMF TYDESTKLFW FNPSSFETEG
UBE3A (ubiquitin-protein ligase E3A, AKA E6AP) is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and transfers it to its substrates. Several substrates have been identified including the ARNTL/BMAL1, ARC, RAD23A, RAD23B, MCM7, annexin A1, the PML tumor suppressor, and the cell cycle regulator CDKN1B. Additionally, UBE3A may function as a cellular quality control ubiquitin ligase by helping the degradation of the cytoplasmic misfolded proteins. UBE3A also plays an important role in the regulation of the circadian clock. Defects in UBE3A are a cause of Angelman syndrome (AS).
Specifications
| E6AP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q05086 | |
| UBE3A | |
| Recombinant human UBE3A (immunogen range is 181-610). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K29054_11H8 | |
| Unconjugated | |
| UBE3A | |
| 4732496B02; 5830462N02Rik; A130086L21Rik; ANCR; AS; CTCL tumor antigen se37-2; E6AP; E6-AP; E6-AP ubiquitin protein ligase; E6AP ubiquitin-protein ligase; EPVE6AP; FLJ26981; HECT-type ubiquitin transferase E3A; HPVE6A; human papilloma virus E6-associated protein; human papillomavirus E6-associated protein; mKIAA4216; oncogenic protein-associated protein E6-AP; Renal carcinoma antigen NY-REN-54; UBE3A; ubiquitin conjugating enzyme E3A; ubiquitin protein ligase E3A; ubiquitin-protein ligase E3A | |
| Mouse | |
| Protein A | |
| RUO | |
| 7337 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.