missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | E2F8 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16861634
|
Proteintech
13425-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16851634
|
Proteintech
13425-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E2F transcription factors, such as E2F8, are essential for orchestrating expression of genes required for cell cycle progression and proliferation.Specifications
| E2F8 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| A0AVK6, F1LMN3, Q58FA4 | |
| 108961, 308607, 79733 | |
| E2F8 Fusion Protein Ag4216 | |
| Primary | |
| -20°C | |
| E2F8 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E2F 8, E2F transcription factor 8, E2F8, Transcription factor E2F8 | |
| E2f8 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title