missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13425-1-AP-20UL
This item is not returnable.
View return policy
Description
E2F transcription factors, such as E2F8, are essential for orchestrating expression of genes required for cell cycle progression and proliferation.Specifications
| E2F8 | |
| Polyclonal | |
| Unconjugated | |
| E2F8 | |
| E2F 8, E2F transcription factor 8, E2F8, Transcription factor E2F8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108961, 308607, 79733 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A0AVK6, F1LMN3, Q58FA4 | |
| E2f8 | |
| E2F8 Fusion Protein Ag4216 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction