missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F2 Recombinant Rabbit Monoclonal Antibody (23GB5250), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | E2F2 |
|---|---|
| Clone | 23GB5250 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089383
|
Invitrogen™
MA552822 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088084
|
Invitrogen™
MA552823 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E2F2 Recombinant Monoclonal antibody specifically detects E2F2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| E2F2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P56931, Q14209 | |
| 1870, 242705, 684111 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5250 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| E2F transcription factor 2; E2F2; E2F-2; LOW QUALITY PROTEIN: transcription factor E2F2; transcription factor E2F2 | |
| A synthesized peptide derived from human E2F2 (313-375AA). | |
| Primary | |
| -20°C | |
| E2F2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title