missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ E.E. Broth (Buffered Glucose Brilliant Green Bile Broth)

Product Code. 13295799
Change view
Click to view available options
Description:
E.E. Broth (Buffered Glucose Brilliant Green Bile Broth)
Quantity:
50 Tubes/Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Unit Size
13295799 E.E. Broth (Buffered Glucose Brilliant Green Bile Broth) 50 Tubes/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13295799 Supplier Thermo Scientific™ Supplier No. TV5041E
  • ONLINE EXCLUSIVE PRICE
    £98.50
    £66.51 / Pack of 50
    Save £31.99
    32% Off
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enrich Enterobacteriaceae in convenient ready-prepared tubes for the bacteriological examination of foods.

Examine the sanitary quality of food samples easily with Thermo Scientific™ E.E. Broth (Buffered Glucose Brilliant Green Bile Broth). Enrich Enterobacteriaceae in convenient, ready-prepared tubes for the bacteriological examination of foods.

To monitor the sanitary quality of food and drugs, it is important to enumerate Enterobacteriaceae in samples accurately. E.E. Broth is recommended as an enrichment medium for Enterobacteriaceae in the bacteriological examination of foods and animal feed stuffs.

  • Improved inhibition: non-Enterobacteriaceae are inhibited due to brilliant green and bile salts in the medium
  • Improved reliability: overcomes inhibition of small numbers of Enterobacteriaceae cells due to bile salt variations
  • Improved accuracy: the entire Enterobacteriaceae group can be used as indicator organisms avoiding discrepancies which can occur with standard coli-aerogenes test

The medium is more inhibitory to non-Enterobacteriaceae than other non-selective media (e.g., Mannitol broth or Lactose broth) due to the presence of brilliant green and bile salts.

The reliability of methods used to enumerate Enterobacteriaceae depends on resuscitation of damaged cells. Weakened cells arise from exposure to dehydration, low pH and other unfavourable conditions. E.E. Broth's formulation includes purified ox bile to overcome inhibition of small numbers of Enterobacteriaceae cells due to bile salt variations. A preliminary assay can be used to check growth by inoculating approximately one viable cell per medium unit.

For the bacteriological evaluation of processed foods, the entire Enterobacteriaceae group can be used as indicator organisms. This overcomes discrepancies that can arise when lactose-negative, anaerogenic lactose-positive or late lactose fermenting enterobacteria are present but are missed by the standard coli-aerogenes tests.

To overcome these problems, lactose media have been replaced by those containing glucose. Mossel et al1 cited several examples which referred to foods contaminated with salmonellae, although results for coliforms were negative. A later example quoted by Mossel2 involved an outbreak of diarrhea caused by French mold-fermented soft cheese contaminated by Escherichia coli serotype O124. This organism was lactose-negative and, therefore, was not detected in coliform tests, but only recognised when the commodity was tested for Enterobacteriaceae since it fermented glucose rapidly.

E.E. Broth should be used as an enrichment broth in conjunction with Violet Red Bile Glucose Agar (Part No. PO5043A). When specific organisms are required, rather than Enterobacteriaceae in general, subcultures must be made onto lactose differential media, e.g., Desoxycholate Citrate Agar.

Not all products are available for sale in all territories. Please inquire.

TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage 2 to 25°C
Description E.E. Broth (Buffered Glucose Brilliant Green Bile Broth)
Format Tubes
Product Type Microbiology Media
Fill Weight 10.0mL + 0.5mL
Packaging Type Wrapped in single cellulose-based film wrap
Quantity 50 Tubes/Pk.
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.