Learn More
Corning™ E-Cube Fittings Kit
Product Code. 10688829 Shop All Corning Products
Click to view available options
Product Type:
Fittings
Unit Size:
Each
Product Code. 10688829
Brand: Corning™ 3287
Only null left
Add to basket
Add to basket
3/16 inch Norprene™ Pump tubing (18 inches)
E-Cube™ Fittings Kit contains:
  • 3/16 inch Norprene™ Pump tubing (18 inches)
  • Tubing ratchet clamps (6ea)
  • Barbed T-fittings (2ea)
  • Barbed Reducing Connector (1ea)
  • 4 inch Wire ties (20ea)
  • Barbed Y-connector (1ea)
  • Barbed 50mm PTFE syringe filters (2ea)
  • Male barbed luer fittings (5ea)
  • Female barbed luer fittings (5ea)
  • Male luer plugs (2ea)
  • Interlink injection sites (2ea)
  • Blunt plastic cannulas (1 sleeve)
  • 1.0 inch I.D. x 1.25 inch O.D., 1/8 inch thickness O-rings (3ea)
TRUSTED_SUSTAINABILITY
Specifications
Includes 3/16 in. Norprene Pump tubing, Tubing ratchet clamps, Barbed T-fittings, Barbed Reducing Connector, 4 inch Wire ties, Barbed Y-connector, Barbed 50mm PTFE syringe filters, Male barbed luer fittings, Female barbed luer fittings, Male luer plugs, Interlink
No. per Case 1
Product Type Fittings
Quantity 1Pack