missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E. Coli J5 LPS Antibody (2D7/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£170.00 - £366.00
Specifications
| Antigen | E. Coli J5 LPS |
|---|---|
| Clone | 2D7/1 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18032226
|
Novus Biologicals
NB100-64543-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18735523
|
Novus Biologicals
NB100-64543 |
0.1 mg |
£366.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E. Coli J5 LPS Monoclonal specifically detects E. Coli J5 LPS in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
| E. Coli J5 LPS | |
| 0.5 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| E. coli J5 cells. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2D7/1 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Escherichia coli | |
| IgG2b | |
| Protein A purified | |
| This is specific for E. coli J5 LPS. It has also been found to react with K. pneumoniae, S. sonnei and S. typhimurium. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title