missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E. Coli J5 LPS Antibody (2D7/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64543
This item is not returnable.
View return policy
Description
E. Coli J5 LPS Monoclonal specifically detects E. Coli J5 LPS in Bacteria samples. It is validated for Western Blot, ELISA.
Specifications
| E. Coli J5 LPS | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| This is specific for E. coli J5 LPS. It has also been found to react with K. pneumoniae, S. sonnei and S. typhimurium. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, ELISA | |
| 2D7/1 | |
| Unconjugated | |
| Escherichia coli | |
| E. coli J5 cells. | |
| 0.1 mg | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction