missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin Antibody (CDH1/2208R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07902-100ug
This item is not returnable.
View return policy
Description
E-Cadherin Monoclonal antibody specifically detects E-Cadherin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
| E-Cadherin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, Immunofluorescence 1-2ug/ml | |
| P12830, P12830 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| CDH1/2208R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
| Recombinant full-length human E-Cadherin protein (Uniprot: P12830) | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| 999 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction