missing translation for 'onlineSavingsMsg'
Learn More

(E)-1-(3,4-Dimethoxyphenl)butadiene (DMPBD), TRC

Product Code. 30387898
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387898 10 mg -
30374552 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387898 Supplier Toronto Research Chemicals Supplier No. TRCD75560510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75560-74-6
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
Synonym (E)-1-(3,4-Dimethoxyphenyl)buta-1,3-diene, 4-(1E)-1,3-Butadien-1-yl-1,2-dimethoxybenzene, 4-(1E)-1,3-butadienyl-1,2-dimethoxy- (9CI)-benzene
SMILES COc1ccc(\C=C\C=C)cc1OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H14O2/c1-4-5-6-10-7-8-11(13-2)12(9-10)14-3/h4-9H,1H2,2-3H3/b6-5+
Formula Weight 190.0994
Chemical Name or Material (E)-1-(3,4-Dimethoxyphenl)butadiene (DMPBD)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.