CAS RN 75560-74-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(E)-1-(3,4-Dimethoxyphenl)butadiene (DMPBD), TRC
CAS: 75560-74-6 Molecular Formula: C12H14O2 Molecular Weight (g/mol): 190.24 Synonym: (E)-1-(3,4-Dimethoxyphenyl)buta-1,3-diene,4-(1E)-1,3-Butadien-1-yl-1,2-dimethoxybenzene,4-(1E)-1,3-butadienyl-1,2-dimethoxy- (9CI)-benzene SMILES: COc1ccc(\C=C\C=C)cc1OC